Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphorylcholine-d4 | 326495-35-6 | 99.0% | 794.17 g/mol | C44H84D4NO8P | 1 MG
SDP

Catalog No. 5000209560
Encompass_Preferred

DSPC-d4 (1,2-distearoyl-sn-glycero-3-phosphorylcholine-d4) is the deuterium-labeled form of DSPC used as an internal standard and tracer in analytical methods, and as a structural lipid for liposome and lipid nanoparticle formulations.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS
  • Used as a lipid component in liposome and LNP systems
  • Molecular formula C44H84D4NO8P and molecular weight 794.17 g/mol
  • Purity: 99.0%
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-09560 Supplier Medchemexpress LLC Supplier No. HYW040193S41MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPC-d4 (1,2-distearoyl-sn-glycero-3-phosphorylcholine-d4) is the deuterium-labeled form of DSPC used as an internal standard and tracer in analytical methods, and as a structural lipid for liposome and lipid nanoparticle formulations.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS
  • Used as a lipid component in liposome and LNP systems
  • Molecular formula C44H84D4NO8P and molecular weight 794.17 g/mol
  • Purity: 99.0%
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.