Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphorylcholine-d70 | 56952-01-3 | 99.0% | 860.58 | C44H18D70NO8P | 10 MG
SDP

Catalog No. 5000208772
Encompass_Preferred

DSPC-d70 is a deuterium-labeled 1,2-distearoyl-sn-glycero-3-phosphorylcholine supplied as a high-purity research reagent. It is used as a tracer or internal standard for quantitative lipid analysis by NMR, GC-MS, or LC-MS, and as a lipid component in liposome and lipid nanoparticle formulations.

  • Deuterated (d70) phosphatidylcholine for analytical applications.
  • High purity (99.0%) suitable for quantitative assays.
  • Molecular weight 860.58; formula C44H18D70NO8P.
  • CAS number 56952-01-3 for unambiguous identification.
  • Available as a 10 mg pack for laboratory use.
  • Intended for research use only.

Catalog No. 50-002-08772 Supplier Medchemexpress LLC Supplier No. HYW040193S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPC-d70 is a deuterium-labeled 1,2-distearoyl-sn-glycero-3-phosphorylcholine supplied as a high-purity research reagent. It is used as a tracer or internal standard for quantitative lipid analysis by NMR, GC-MS, or LC-MS, and as a lipid component in liposome and lipid nanoparticle formulations.

  • Deuterated (d70) phosphatidylcholine for analytical applications.
  • High purity (99.0%) suitable for quantitative assays.
  • Molecular weight 860.58; formula C44H18D70NO8P.
  • CAS number 56952-01-3 for unambiguous identification.
  • Available as a 10 mg pack for laboratory use.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.