Learn More
Medchemexpress LLC 1,2-distearoyl-sn-glycero-3-phosphorylcholine-d79 | 326495-39-0 | 99.3% | 869.63 g/mol | C44H9D79NO8P | 1 MG
SDP
Catalog No. 5000208662
Encompass_Preferred
Deuterated form of 1,2-distearoyl-sn-glycero-3-phosphorylcholine used as a saturated phospholipid for liposome and lipid nanoparticle formulations. Deuteration replaces hydrogen atoms with deuterium to enable analytical tracing and may affect metabolic stability.
  • Deuterium-labeled phospholipid for tracing and mass-shift studies.
  • High purity suitable for formulation and analytical work.
  • Small pack sizes for research-scale experiments.
  • Compatible with liposome and lipid nanoparticle synthesis protocols.
Catalog No. 50-002-08662 Supplier Medchemexpress LLC Supplier No. HYW040193S21MG
Deuterated form of 1,2-distearoyl-sn-glycero-3-phosphorylcholine used as a saturated phospholipid for liposome and lipid nanoparticle formulations. Deuteration replaces hydrogen atoms with deuterium to enable analytical tracing and may affect metabolic stability.
  • Deuterium-labeled phospholipid for tracing and mass-shift studies.
  • High purity suitable for formulation and analytical work.
  • Small pack sizes for research-scale experiments.
  • Compatible with liposome and lipid nanoparticle synthesis protocols.