Learn More
Medchemexpress LLC 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) | 1069-79-0 | 100.0% | 748.07 | 500 MG
SDP
Catalog No. 5000438454
Encompass_Preferred
1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphoethanolamine (PE) lipid used in the synthesis of liposomes. It cannot sustain a lamellar form at pH values below approximately 8.5, which is the pKa of the ethanolamine moiety. This product is for research use only.
  • Phosphoethanolamine (PE) lipid
  • Used in the synthesis of liposomes
  • Cannot sustain lamellar form at pH values below approximately 8.5
  • For research use only
Catalog No. 50-004-38454 Supplier Medchemexpress LLC Supplier No. HY112530500MG
1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphoethanolamine (PE) lipid used in the synthesis of liposomes. It cannot sustain a lamellar form at pH values below approximately 8.5, which is the pKa of the ethanolamine moiety. This product is for research use only.
  • Phosphoethanolamine (PE) lipid
  • Used in the synthesis of liposomes
  • Cannot sustain lamellar form at pH values below approximately 8.5
  • For research use only