Learn More
Medchemexpress LLC 1,2-Distearoyl-sn-glycerol | 10567-21-2 | 98.0% | 625.02 | 100 MG
SDP
Catalog No. 5000438158
Encompass_Preferred
1,2-Distearoyl-sn-glycerol (DSG) is a glyceride compound. It functions as an internal standard for the analysis of molecular species of diacylglycerol (DAG) and other phospholipids in rat brain.
  • Glyceride compound
  • Functions as an internal standard
  • Used for analysis of molecular species of diacylglycerol (DAG)
  • Used for analysis of other phospholipids in rat brain
Catalog No. 50-004-38158 Supplier Medchemexpress LLC Supplier No. HY112552100MG
1,2-Distearoyl-sn-glycerol (DSG) is a glyceride compound. It functions as an internal standard for the analysis of molecular species of diacylglycerol (DAG) and other phospholipids in rat brain.
  • Glyceride compound
  • Functions as an internal standard
  • Used for analysis of molecular species of diacylglycerol (DAG)
  • Used for analysis of other phospholipids in rat brain