Learn More
Medchemexpress LLC 1,2-Distearoyl-sn-glycerol | 10567-21-2 | 98.0% | 625.02 | 250 MG
SDP
Catalog No. 5000439032
Encompass_Preferred
1,2-Distearoyl-sn-glycerol (DSG) is a glyceride compound. It can be used as an internal standard for the analysis of molecular species of diacylglycerol (DAG) and other phospholipids in rat brain.
  • Used as an internal standard for the analysis of molecular species of diacylglycerol (DAG) and other phospholipids in rat brain.
Catalog No. 50-004-39032 Supplier Medchemexpress LLC Supplier No. 250MGHY112552
1,2-Distearoyl-sn-glycerol (DSG) is a glyceride compound. It can be used as an internal standard for the analysis of molecular species of diacylglycerol (DAG) and other phospholipids in rat brain.
  • Used as an internal standard for the analysis of molecular species of diacylglycerol (DAG) and other phospholipids in rat brain.