Learn More
Apexbio Technology LLC 1-2-Distearoyl-sn-glycerol
SDP
Catalog No. 5000568560
Encompass_Preferred
1 2-Distearoyl-sn-glycerol (CAS No 10567-21-2) is a diacylglycerol utilized in biomedical research particularly in the study of lipid signaling pathways As a glycerolipid it plays a vital role in the activation of protein kinase C (PKC) which is crucial for various cellular processes including cell growth and apoptosis Common in vitro activities include modulation of PKC in the nanomolar to low micromolar range It is frequently used in cell models to investigate lipid metabolism and signal transduction In animal studies it aids in understanding lipid-related disorders and drug screening Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68560 Supplier Apexbio Technology LLC Supplier No. C4877250
1 2-Distearoyl-sn-glycerol (CAS No 10567-21-2) is a diacylglycerol utilized in biomedical research particularly in the study of lipid signaling pathways As a glycerolipid it plays a vital role in the activation of protein kinase C (PKC) which is crucial for various cellular processes including cell growth and apoptosis Common in vitro activities include modulation of PKC in the nanomolar to low micromolar range It is frequently used in cell models to investigate lipid metabolism and signal transduction In animal studies it aids in understanding lipid-related disorders and drug screening Experimental concentrations typically depend on the specific research design