Learn More
Medchemexpress LLC 1,2-Dithiolane-3-pentanoic acid, 2,5-dioxo-1-pyrrolidinyl ester | 40846-94-4 | 98.0% | C12H17NO4S2 | 1 G
SDP
Catalog No. 5000353855
Encompass_Preferred
α-Lipoic acid-NHS is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. They achieve this by connecting two ligands with a linker: one targeting an E3 ubiquitin ligase and the other binding to the protein of interest. This product is for research use only.
  • Alkyl/ether-based linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53855 Supplier Medchemexpress LLC Supplier No. HY1413361G
α-Lipoic acid-NHS is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. They achieve this by connecting two ligands with a linker: one targeting an E3 ubiquitin ligase and the other binding to the protein of interest. This product is for research use only.
  • Alkyl/ether-based linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins