Learn More
Grainger 1/2 DOM BLK XH N/R T/T COUPLING
Catalog No. 502751565
Encompass_Preferred
The Grainger 14J821 is a 14-inch diameter, 8-inch deep, 21-gallon capacity plastic pail with lid. It is made from high density polyethylene and has a wire bail handle for easy lifting and carrying. The pail has graduated volume markings up to 21 gallons. It can be used for a variety of applications including mixing, storing, and transporting liquids and dry goods. The snap-on lid helps keep contents secure.
Catalog No. 50-275-1565 Supplier Grainger Supplier No. 14J821
The Grainger 14J821 is a 14-inch diameter, 8-inch deep, 21-gallon capacity plastic pail with lid. It is made from high density polyethylene and has a wire bail handle for easy lifting and carrying. The pail has graduated volume markings up to 21 gallons. It can be used for a variety of applications including mixing, storing, and transporting liquids and dry goods. The snap-on lid helps keep contents secure.