Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Epoxybutane

Catalog No. 109975250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-997-5250ML 250 mL
10-997-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-997-5250ML Supplier Sigma Aldrich Supplier No. 109975250ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Epoxybutane

Specifications

CAS 106-88-7
Density 0.829 g/mL (at 25°C)
Boiling Point 63°C
Molecular Formula C4H8O
Refractive Index n20/D 1.384 (literature)
Linear Formula C4H8O
Quantity 250 mL
MDL Number MFCD00005153
Synonym alpha-Butylene oxide; 1,2-Butylene oxide; 1-Butene oxide; Ethyloxirane
Molecular Weight (g/mol) 72.11
Percent Purity 99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.