Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ (++)-1,2-Epoxybutane, 99%

Catalog No. AAH3658122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3658122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3658122 Supplier Thermo Scientific™ Supplier No. H3658122
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 106-88-7 | C4H8O | 72.11 g/mol

TRUSTED_SUSTAINABILITY

Specifications

CAS 106-88-7
Molecular Formula C4H8O
Quantity 100 g
MDL Number MFCD00005153
UN Number UN3022
Synonym α-Butylene oxide;Ethyloxirane
InChI Key RBACIKXCRWGCBB-UHFFFAOYNA-N
SMILES CCC1CO1
IUPAC Name 2-ethyloxirane
Molecular Weight (g/mol) 72.11
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Show More Show Less
ShelfLife 10 Years
EINECSNumber 203-438-2
RTECSNumber EK3675000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.