Learn More
Medchemexpress LLC 1,2-epoxydecane | 2404-44-6 | MFCD00005158 | ≥98.0% | 156.27 | C10H20O | 25 G
SDP
Catalog No. 5000283734
Encompass_Preferred
1,2-Epoxydecane (2-octyloxirane) is an organic epoxide reagent used in chemical and life-science research for alkylation and ring-opening reactions. It is supplied as a high-purity, colorless to light-yellow liquid (formula C10H20O, MW 156.27) and should be stored under recommended cold conditions to maximize shelf life.
  • Epoxide functionality suitable for alkylation and ring-opening chemistry.
  • High purity (≥98.0%) for research applications.
  • Liquid, colorless to light yellow, easy to handle and dispense.
  • Molecular formula C10H20O and molecular weight 156.27 g·mol-1.
  • Available in laboratory pack sizes (for example, 25 g) for small-scale use.
  • Stable when stored at recommended cold temperatures, aiding long-term storage.
Catalog No. 50-002-83734 Supplier Medchemexpress LLC Supplier No. HYW14270225G
1,2-Epoxydecane (2-octyloxirane) is an organic epoxide reagent used in chemical and life-science research for alkylation and ring-opening reactions. It is supplied as a high-purity, colorless to light-yellow liquid (formula C10H20O, MW 156.27) and should be stored under recommended cold conditions to maximize shelf life.
  • Epoxide functionality suitable for alkylation and ring-opening chemistry.
  • High purity (≥98.0%) for research applications.
  • Liquid, colorless to light yellow, easy to handle and dispense.
  • Molecular formula C10H20O and molecular weight 156.27 g·mol-1.
  • Available in laboratory pack sizes (for example, 25 g) for small-scale use.
  • Stable when stored at recommended cold temperatures, aiding long-term storage.