Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxyhexadecane 80.0+%, TCI America™
SDP

Catalog No. E031625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E031625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E031625ML Supplier TCI America Supplier No. E031625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7320-37-8
Molecular Formula C16H32O
Molecular Weight (g/mol) 240.43
MDL Number MFCD00005151
InChI Key DSZTYVZOIUIIGA-UHFFFAOYNA-N
Synonym 1,2-Hexadecylene Oxide
PubChem CID 23741
ChEBI CHEBI:34054
IUPAC Name 2-tetradecyloxirane
SMILES CCCCCCCCCCCCCCC1CO1

Specifications

Melting Point 22°C
Color White
Boiling Point 180°C
Quantity 25 mL
Formula Weight 240.43
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Epoxyhexadecane
EINECSNumber (2)-0220
RTECSNumber ML9450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.