Learn More
Medchemexpress LLC 1,2-Ethanediamine, N1-[[2-(2,3-dichlorophenoxy)phenyl]methyl]-N1,N2-dimethyl- (hydrochloride) | 2289726-31-2 | 99.9% | 375.72 | 25 MG
SDP
Catalog No. 5000391392
Encompass_Preferred
DCPT1061 hydrochloride is a compound with a strong inhibitory effect on PRMT1, PRMT6, and PRMT8 in vitro. It shows little inhibitory effect on epigenetic enzymes such as PRMT3, PRMT4, and PRMT5. This compound also exhibits antitumor effects.
  • Strong inhibitor of PRMT1, PRMT6, and PRMT8
  • Exhibits antitumor effects
  • Relevant for cancer targeted therapy
  • Applicable in cancer metabolism and metastasis research
  • Used in epigenetics research, specifically histone methyltransferase studies
  • Useful for research in blood or cardio-cerebrovascular disease, blood disease, leukemia/lymphoma/myeloma
Catalog No. 50-003-91392 Supplier Medchemexpress LLC Supplier No. 25MGHY162003A
DCPT1061 hydrochloride is a compound with a strong inhibitory effect on PRMT1, PRMT6, and PRMT8 in vitro. It shows little inhibitory effect on epigenetic enzymes such as PRMT3, PRMT4, and PRMT5. This compound also exhibits antitumor effects.
  • Strong inhibitor of PRMT1, PRMT6, and PRMT8
  • Exhibits antitumor effects
  • Relevant for cancer targeted therapy
  • Applicable in cancer metabolism and metastasis research
  • Used in epigenetics research, specifically histone methyltransferase studies
  • Useful for research in blood or cardio-cerebrovascular disease, blood disease, leukemia/lymphoma/myeloma