Learn More
Medchemexpress LLC 1,2-Ethanediamine, N1-[[3-[4,4-bis(ethoxymethyl)cyclohexyl]-1H-pyrazol-4-yl]methyl]-N1,N2-dimethyl | 1629013-22-4 | 99.4% | 366.54 g/mol | C20H38N4O2 | 5 MG
SDP
Catalog No. 5000262265
Encompass_Preferred
GSK3368715 (EPZ019997) is an orally active, reversible, S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferase (PRMT) inhibitor used for biochemical and cell-based research into arginine methylation and oncology-related pathways.
  • Orally active, reversible type I PRMT inhibitor.
  • SAM uncompetitive mechanism of action.
  • Potent activity with low-nanomolar activity against PRMT1 and PRMT8.
  • High purity (99.4% by HPLC) for research applications.
  • Molecular formula C20H38N4O2; molecular weight 366.54 g/mol.
  • Intended for in vitro and cellular research use only.
  • Supplied in small research pack sizes suitable for screening and assays.
Catalog No. 50-002-62265 Supplier Medchemexpress LLC Supplier No. HY1287175MG
GSK3368715 (EPZ019997) is an orally active, reversible, S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferase (PRMT) inhibitor used for biochemical and cell-based research into arginine methylation and oncology-related pathways.
  • Orally active, reversible type I PRMT inhibitor.
  • SAM uncompetitive mechanism of action.
  • Potent activity with low-nanomolar activity against PRMT1 and PRMT8.
  • High purity (99.4% by HPLC) for research applications.
  • Molecular formula C20H38N4O2; molecular weight 366.54 g/mol.
  • Intended for in vitro and cellular research use only.
  • Supplied in small research pack sizes suitable for screening and assays.