Learn More
Sigma Aldrich 1-[2-(Fluorolsulfonyl)ethyl]-L-proline
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 5-methyl-1-hexanol, an aliphatic alcohol, can be prepared by the reduction of 5-methylhexanoic acid. it is predicted to have a fruity odor based on fuzzy partition and self organising maps (som) analysis data. 5-methyl-1-hexanol is a volatile organic compound found in:
- Alstonia boonei leaves
- 'hayward' and 'hort16a' kiwifruit
- Tuber melanosporum fruiting body
Specifications
Specifications
| Molecular Formula | C7H5FO4S |
| Linear Formula | C7H5FO4S |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 204.18 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.