Learn More
Accuflow Systems Saint-Gobain Mini Adapter, Polypropylene, 1/2 in FPT, 3/4 in Tri-Clamp
Catalog No. NC3232620
Encompass_Preferred
This polypropylene mini adapter provides a sanitary transition between a 1/2 in female pipe thread (FPT) and a 3/4 in mini tri-clamp connection for high-purity process lines. It is designed for cleanability and minimal fluid entrapment in applications that require sanitary, smooth-bore fittings.
  • Material: Polypropylene
  • Connections: 1/2 in female pipe thread (FPT) to 3/4 in mini tri-clamp
  • Design: Smooth inner bore for reduced entrapment and improved hydraulics
  • Cleaning: Compatible with clean-in-place (CIP) procedures
  • Applications: Pharmaceutical, biotechnology, DI water, semiconductor, food and beverage
  • Standards: Tubing family supports FDA, USDA, 3A, USP Class VI, and NSF requirements
Catalog No. NC3232620 Supplier Accuflow Systems Supplier No. TAD07508F
This polypropylene mini adapter provides a sanitary transition between a 1/2 in female pipe thread (FPT) and a 3/4 in mini tri-clamp connection for high-purity process lines. It is designed for cleanability and minimal fluid entrapment in applications that require sanitary, smooth-bore fittings.
  • Material: Polypropylene
  • Connections: 1/2 in female pipe thread (FPT) to 3/4 in mini tri-clamp
  • Design: Smooth inner bore for reduced entrapment and improved hydraulics
  • Cleaning: Compatible with clean-in-place (CIP) procedures
  • Applications: Pharmaceutical, biotechnology, DI water, semiconductor, food and beverage
  • Standards: Tubing family supports FDA, USDA, 3A, USP Class VI, and NSF requirements