Learn More
Chemglass Life Sciences Graded Seal, 1/2in OD Kovar to Borosilicate
SDP
Catalog No. CG30606
Encompass_Preferred
Graded Seal, 1/2in OD Kovar to Borosilicate. For connection of borosilicate glass components to metal. Kovar length is approximately 25mm on all sizes, while the borosilicate tubing is approximately 100mm.
Catalog No. CG30606 Supplier Chemglass Life Sciences Supplier No. CG30606
Graded Seal, 1/2in OD Kovar to Borosilicate. For connection of borosilicate glass components to metal. Kovar length is approximately 25mm on all sizes, while the borosilicate tubing is approximately 100mm.