Learn More
COLDER PRODUCTS COMPANY DC Body AseptiQuik CPC 1/2 Hose Barb Polycarb White w/blue pull tab Free floating Seal Silicone platinum-cured USP Class VI ADCF Gamma sterilized Temp Range 4 deg to 40 deg C Pressure Up to 60psi
Catalog No. 503131284
Encompass_Preferred
DC Body AseptiQuik CPC 1/2 Hose Barb Polycarb White w/blue pull tab Free floating Seal Silicone platinum-cured USP Class VI ADCF Gamma sterilized Temp Range 4 deg to 40 deg C Pressure Up to 60psi
Catalog No. 50-313-1284 Supplier COLDER PRODUCTS COMPANY Supplier No. AQCDC17008
DC Body AseptiQuik CPC 1/2 Hose Barb Polycarb White w/blue pull tab Free floating Seal Silicone platinum-cured USP Class VI ADCF Gamma sterilized Temp Range 4 deg to 40 deg C Pressure Up to 60psi