Learn More
Medchemexpress LLC 1-(2-Hydroxy-4,6-dimethoxyphenyl)-4-(piperazin-1-yl)butan-1-one dihydrochloride | 99.9% | 381.29 | 100 MG
SDP
Catalog No. 5000438676
Encompass_Preferred
Y13g dihydrochloride is a potent inhibitor of both acetylcholinesterase (AChE) and interleukin-6 (IL-6), which are significant targets in Alzheimer's Disease progression. This compound has been shown to reverse STZ-induced memory deficits and demonstrate histopathology similar to normal animal models. It is intended for research use only.
  • Potent inhibitor of AChE and IL-6
  • Reverses STZ-induced memory deficit
  • Shows histopathology similar to normal animals
  • Suitable for neurological disease research
Catalog No. 50-004-38676 Supplier Medchemexpress LLC Supplier No. HY115910A100MG
Y13g dihydrochloride is a potent inhibitor of both acetylcholinesterase (AChE) and interleukin-6 (IL-6), which are significant targets in Alzheimer's Disease progression. This compound has been shown to reverse STZ-induced memory deficits and demonstrate histopathology similar to normal animal models. It is intended for research use only.
  • Potent inhibitor of AChE and IL-6
  • Reverses STZ-induced memory deficit
  • Shows histopathology similar to normal animals
  • Suitable for neurological disease research