Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(2-Hydroxy-4,6-dimethoxyphenyl)-4-(piperazin-1-yl)butan-1-one dihydrochloride | 99.9% | 381.29 | 50 MG
SDP

Catalog No. 5000435722
Encompass_Preferred

Y13g dihydrochloride is a chemical compound supplied by MedChemExpress. It has a molecular weight of 381.29 and a purity of 99.89% as determined by LCMS. It appears as an off-white to light yellow solid and is stored at -20°C in sealed conditions away from moisture.

  • Appears as an off-white to light yellow solid
  • Purity of 99.89% verified by LCMS
  • Consistent with ¹H NMR spectrum and LCMS analysis
  • Recommended storage at -20°C in sealed conditions away from moisture

Catalog No. 50-004-35722 Supplier Medchemexpress LLC Supplier No. HY115910A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Y13g dihydrochloride is a chemical compound supplied by MedChemExpress. It has a molecular weight of 381.29 and a purity of 99.89% as determined by LCMS. It appears as an off-white to light yellow solid and is stored at -20°C in sealed conditions away from moisture.

  • Appears as an off-white to light yellow solid
  • Purity of 99.89% verified by LCMS
  • Consistent with ¹H NMR spectrum and LCMS analysis
  • Recommended storage at -20°C in sealed conditions away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.