Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxy-4-methylphenyl)ethan-1-one, 97%, Thermo Scientific™

Catalog No. BTB09999DE
Change view
Click to view available options
5g; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BTB09999DE 5g; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09999DE Supplier Thermo Scientific Chemicals Supplier No. BTB09999DE

missing translation for 'validMainframeMsg'

Specifications

CAS 6921-64-8
CAS Min % 96.5
CAS Max % 100.0
Density 1.0800g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H10O2
Quantity 5g
Synonym 1-2-hydroxy-4-methylphenyl ethanone, 1-2-hydroxy-4-methylphenyl ethan-1-one, 2'-hydroxy-4'-methylacetophenone, ethanone, 1-2-hydroxy-4-methylphenyl, 2-hydroxy-4-methylacetophenone, 4-methyl-2-hydroxyacetophenone, 1-2-hydroxy-4-methyl-phenyl-ethanone, unii-q168p7rg3b, 1-acetyl-2-hydroxy-4-methylbenzene
InChI Key LYKDOWJROLHYOT-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)C(=O)C)O
IUPAC Name 1-(2-hydroxy-4-methylphenyl)ethanone
Molecular Weight (g/mol) 150.177
PubChem CID 81338
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 1-(2-Hydroxy-4-methylphenyl)ethan-1-one
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.