Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxyethyl)-3-methylimidazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. H12545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12545G 5 g
H125425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12545G Supplier TCI America Supplier No. H12545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61755-34-8
Molecular Formula C6H11ClN2O
MDL Number MFCD12761437

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Hydroxyethyl)-3-methylimidazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.