Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxyethyl)-3-methylimidazolium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. H14925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14925G 5 g
H149225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14925G Supplier TCI America Supplier No. H14925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 374564-83-7
Molecular Formula C6H11BF4N2O
Molecular Weight (g/mol) 213.971
MDL Number MFCD12761438
InChI Key KLTUZFZUYLXTFF-UHFFFAOYSA-N
PubChem CID 11344909
IUPAC Name 2-(3-methylimidazol-3-ium-1-yl)ethanol;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C[N+]1=CN(C=C1)CCO

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 213.97
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-(2-Hydroxyethyl)-3-methylimidazolium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.