Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxyethyl)ethyleneimine 97.0+%, TCI America™
SDP

Catalog No. H056725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H056725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H056725ML Supplier TCI America Supplier No. H056725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1072-52-2
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00005119
InChI Key VYONOYYDEFODAJ-UHFFFAOYSA-N
Synonym 1-aziridineethanol, 2-aziridin-1-yl ethanol, 2-1-aziridinyl ethanol, 1-azirdineethanol, 2-azidin-1-yl ethanol, 1-2-hydroxyethyl ethylenimine, n-2-hydroxyethyl aziridine, ethanol, 2-1-aziridinyl, n-2-hydroxyethyl ethylenimine, n-2-hydroxyethyl ethyleneimine
PubChem CID 14074
ChEBI CHEBI:82393
IUPAC Name 2-(aziridin-1-yl)ethanol
SMILES C1CN1CCO

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 mL
UN Number 2922
Formula Weight 87.12
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Hydroxyethyl)ethyleneimine
EINECSNumber (5)-5559
RTECSNumber CM7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.