Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Hydroxyethyl)piperidine

Catalog No. 116068100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-606-8100G 100 g
11-606-8500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-606-8100G Supplier Sigma Aldrich Supplier No. 116068100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Counter-ion used for investigations of skin permeation of flurbiprofencatalyst used for enantioselective synthesisamine quencher used during synthesis of photoacid generators for arf lithographyadsorbent for capture and release of pressurized carbon dioxide for greenhouse gas controlreagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 3040-44-6
Density 0.973 g/mL (at 25°C (literature))
Boiling Point 199°C to 202°C (lit.)
Molecular Formula C7H15NO
Refractive Index n20/D 1.4804 (literature)
Linear Formula C7H15NO
MDL Number MFCD00006512
Quantity 100 g
Synonym 1-Piperidineethanol; 2-Piperidinoethanol
Molecular Weight (g/mol) 129.2
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.