Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Hydroxyethyl)pyrrole 99.0+%, TCI America™
SDP

Catalog No. H08261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08261G 1 g
H08265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08261G Supplier TCI America Supplier No. H08261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6719-02-4
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00191451
InChI Key ZIOLCZCJJJNOEJ-UHFFFAOYSA-N
PubChem CID 10975438
IUPAC Name 2-pyrrol-1-ylethanol
SMILES C1=CN(C=C1)CCO

Specifications

Quantity 1 g
Formula Weight 111.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Hydroxyethyl)pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.