Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Iodoethynyl)-3-methoxybenzene

Catalog No. RNI001061G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RNI001061G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RNI001061G Supplier Sigma Aldrich Supplier No. RNI001061G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,6-dimethylphenyl isocyanate has been used in the preparation of:
  • Derivatized α-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography
  • Tris( 2,6-dimethylphenylimido)methylrhenium (vii)
  • 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea

Specifications

CAS 223758-81-4
Molecular Formula C9H7IO
Linear Formula C9H7IO
MDL Number MFCD26960818
Quantity 5 g
Molecular Weight (g/mol) 258.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.