Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Iodoethynyl)-4-nitrobenzene

Catalog No. RNI000631G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RNI000631G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RNI000631G Supplier Sigma Aldrich Supplier No. RNI000631G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

p-Tolyl isocyanate reacts with human serum albumin to form antigen which helps in detecting IgE antibodies in workers hypersensitive to toluene diisocyanate.p-Tolyl isocyanate was used to study the catalytic role of supported Rhodium(I) complex toward the reductive carbonylation of nitrobenzene in DMF medium.

Specifications

CAS 69116-22-9
Molecular Formula C8H4INO2
Linear Formula C8H4INO2
MDL Number MFCD26142829
Quantity 5 g
Molecular Weight (g/mol) 273.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.