Learn More
Analytical Sales & Services 1.2µL GC Syringe for LEAP/Gerstel CTC PAL GC Autosamplers
Catalog No. 5000804304
Encompass
1.2µL GC Syringe 7701.2KH for LEAP/Gerstel CTC PAL GC Autosamplers.
26 Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH
Catalog No. 50-008-04304 Supplier Analytical Sales & Services Supplier No. 42031854
1.2µL GC Syringe 7701.2KH for LEAP/Gerstel CTC PAL GC Autosamplers.
26 Gauge, 51mm Length, Poinst Style AS
Qty/Pkg: EACH