Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Mesitylenesulfonyl)imidazole 97.0+%, TCI America™
SDP

Catalog No. M06251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M06251G Supplier TCI America Supplier No. M06251G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50257-39-1
Molecular Formula C12H14N2O2S
Molecular Weight (g/mol) 250.32
MDL Number MFCD00005284
InChI Key XFHVQSPNDDOABS-UHFFFAOYSA-N
Synonym 1-mesitylsulfonyl-1h-imidazole, 1-2-mesitylenesulfonyl imidazole, n-mesitylenesulfonyl imidazole, n-mesitylenesulfonylimidazole, 1-2,4,6-trimethylbenzenesulfonyl imidazole, 1-2,4,6-trimethylphenyl sulphonyl-1h-imidazole, 1h-imidazole, 1-2,4,6-trimethylphenyl sulfonyl, 2,4,6-trimethylphenyl sulfonyl imidazole, acmc-209kkw, mesitylenesulfonylimidazole
PubChem CID 99556
IUPAC Name 1-(2,4,6-trimethylbenzenesulfonyl)-1H-imidazole
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=CN=C1

Specifications

Melting Point 98°C
Color White
Quantity 1 g
Formula Weight 250.32
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Mesitylenesulfonyl)imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.