Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1,2-Methanofullerene C60)-61-carboxylic acid

Catalog No. 65884725MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-884-725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-884-725MG Supplier Sigma Aldrich Supplier No. 65884725MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Behavior of the fullerene derivative in non aqueous capillary electrophoresis has been studied.Reagent for preparation of fullerene derivatives. Interaction of C60 fullerence derivative with L-histidine was studied in detail. The quantum chemical calculations were focused on the enantioselective binding between the two. (1,2-Methanofullerene C60)-61-carboxylic acid may find potential application in biomedical and cosmetics.

Specifications

CAS 155116-19-1
Molecular Formula C62H2O2
Linear Formula C62H2O2
MDL Number MFCD00213323
Quantity 25 mg
Molecular Weight (g/mol) 778.68
Percent Purity 97% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.