Learn More
Medchemexpress LLC 1-(2-Methoxyphenyl)piperazine hydrochloride | 5464-78-8 | 99.59% | 228.72 | 25 G
SDP
Catalog No. 5000454120
Encompass_Preferred
1-(2-Methoxyphenyl)piperazine hydrochloride is a key agent intermediate of antipsychotics with high affinity to the serotonin receptors and 5-HT. It can be used to synthesize the intestinal worm-repellent agents Piperazine phosphate and Piperazine citrate, as well as Fluphenazine, dihydrochloride, and Rifampicin.
  • High affinity to the serotonin receptors and 5-HT.
  • Key intermediate for antipsychotics.
  • Can be used to synthesize intestinal worm-repellent agents.
  • Can be used to synthesize other compounds like Fluphenazine and Rifampicin.
Catalog No. 50-004-54120 Supplier Medchemexpress LLC Supplier No. 25GHYW009681
1-(2-Methoxyphenyl)piperazine hydrochloride is a key agent intermediate of antipsychotics with high affinity to the serotonin receptors and 5-HT. It can be used to synthesize the intestinal worm-repellent agents Piperazine phosphate and Piperazine citrate, as well as Fluphenazine, dihydrochloride, and Rifampicin.
  • High affinity to the serotonin receptors and 5-HT.
  • Key intermediate for antipsychotics.
  • Can be used to synthesize intestinal worm-repellent agents.
  • Can be used to synthesize other compounds like Fluphenazine and Rifampicin.