Learn More
Medchemexpress LLC 1-(2-Methoxyphenyl)piperazine hydrochloride | 5464-78-8 | 99.6% | 228.72 | 100 G
SDP
Catalog No. 5000453127
Encompass_Preferred
1-(2-Methoxyphenyl)piperazine hydrochloride is a key agent intermediate of antipsychotics with high affinity to the serotonin receptors and 5-HT. It can be used to synthesize various compounds.
  • Used to synthesize intestinal worm-repellent agents.
  • Can be used for Piperazine phosphate and Piperazine citrate synthesis.
  • Also used in the synthesis of Fluphenazine dihydrochloride and Rifampicin.
Catalog No. 50-004-53127 Supplier Medchemexpress LLC Supplier No. HYW009681100G
1-(2-Methoxyphenyl)piperazine hydrochloride is a key agent intermediate of antipsychotics with high affinity to the serotonin receptors and 5-HT. It can be used to synthesize various compounds.
  • Used to synthesize intestinal worm-repellent agents.
  • Can be used for Piperazine phosphate and Piperazine citrate synthesis.
  • Also used in the synthesis of Fluphenazine dihydrochloride and Rifampicin.