missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1-(2-Methyl-1-benzothien-3-yl)ethanone

Catalog No. CBR011721G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBR011721G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBR011721G Supplier Sigma Aldrich Supplier No. CBR011721G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-(2-Methyl-1-benzothien-3-yl)ethanone

Specifications

CAS 16810-19-8
Molecular Formula C11H10OS
Linear Formula C11H10OS
MDL Number MFCD08059890
Quantity 100 mg
Molecular Weight (g/mol) 190.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.