Learn More
Accela Chembio Inc 1-[2-(methylsulphonyl)phenyl]piperazine | 5g | 313490-26-5 | MFCD08692378 | 97+% | Shelf Life: 2160 Days | Light Sensitive
Supplier:  Accela Chembio Inc SY0044695G
Encompass_Preferred
1-[2-(methylsulphonyl)phenyl]piperazine | 5g | 313490-26-5 | MFCD08692378 | 97+% | Shelf Life: 2160 Days | Light Sensitive
Catalog No. 50-250-490