Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Mini Hose-Barb Connector, Polypropylene, 1/2 in
Catalog No. NC2323801
Encompass_Preferred
Pure-Fit SIB Mini hose-barb connector designed for contamination-sensitive fluid transfer. Its smooth inner-bore design provides a seamless transition between tubing and fittings, minimizing dead volume and reducing the potential for particle or microbial entrapment. Suitable for sanitary and bioprocessing assemblies.
  • Smooth inner bore reduces dead volume and potential sites for contamination.
  • Material: polypropylene.
  • Inner diameter: 1/2 in (12.7 mm).
  • Mini series hose-barb configuration for compact tubing assemblies.
  • Meets USP Class VI and relevant ISO criteria for bioprocess applications.
  • Common applications include sterile fluid transfer and sanitary tubing assemblies.
Catalog No. NC2323801 Supplier Accuflow Systems Supplier No. PFMS500PPAF
Pure-Fit SIB Mini hose-barb connector designed for contamination-sensitive fluid transfer. Its smooth inner-bore design provides a seamless transition between tubing and fittings, minimizing dead volume and reducing the potential for particle or microbial entrapment. Suitable for sanitary and bioprocessing assemblies.
  • Smooth inner bore reduces dead volume and potential sites for contamination.
  • Material: polypropylene.
  • Inner diameter: 1/2 in (12.7 mm).
  • Mini series hose-barb configuration for compact tubing assemblies.
  • Meets USP Class VI and relevant ISO criteria for bioprocess applications.
  • Common applications include sterile fluid transfer and sanitary tubing assemblies.