Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 1.2 mL, Self-Standing, External Thread, Sterile (CGN-4762-002)
SDP
Catalog No. 502733393
Encompass_Preferred
Sterile 1.2 mL cryogenic vial designed for secure storage and retrieval of biological samples in cryogenic conditions, featuring an external thread and screw cap for a reliable seal.
  • Self-standing design for upright storage and easy handling.
  • External thread and screw cap for secure closure.
  • Sterilized by E-beam for contamination control.
  • Made from USP Class VI polypropylene; DNase/RNase free and non-pyrogenic.
  • Autoclavable at 121 °C for added decontamination options.
  • Suitable for storage in the gas phase of liquid nitrogen.
  • Available in nest or racked packaging; typical rack configuration 9 x 9 with 14 racks per case.
Catalog No. 50-273-3393 Supplier Chemglass Life Sciences Supplier No. CGN4762002
Sterile 1.2 mL cryogenic vial designed for secure storage and retrieval of biological samples in cryogenic conditions, featuring an external thread and screw cap for a reliable seal.
  • Self-standing design for upright storage and easy handling.
  • External thread and screw cap for secure closure.
  • Sterilized by E-beam for contamination control.
  • Made from USP Class VI polypropylene; DNase/RNase free and non-pyrogenic.
  • Autoclavable at 121 °C for added decontamination options.
  • Suitable for storage in the gas phase of liquid nitrogen.
  • Available in nest or racked packaging; typical rack configuration 9 x 9 with 14 racks per case.