Learn More
Andwin Scientific 1.2 ML MICRO-DILUTION TUBES. U
SDP
Catalog No. 50819694
Encompass_Preferred
The 1.2 mL micro-dilution tube is a perfect laboratory apparatus for use in serial diluting and mixing or storing of samples. This tube is compatible with the tube racks 1412-1400 and 1412-1410. It is free of RNase, DNase, DNA, PCR inhibitor, and pyrogen. This product comes in a pack of 1000.
Catalog No. 50-819-694 Supplier Andwin Scientific Supplier No. 14121000
The 1.2 mL micro-dilution tube is a perfect laboratory apparatus for use in serial diluting and mixing or storing of samples. This tube is compatible with the tube racks 1412-1400 and 1412-1410. It is free of RNase, DNase, DNA, PCR inhibitor, and pyrogen. This product comes in a pack of 1000.