Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Nitro-phenyl)-piperazine

Catalog No. MNO0000081G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MNO0000081G 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MNO0000081G Supplier Sigma Aldrich Supplier No. MNO0000081G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(trimethylsiloxy)-1,3-cyclohexadiene ((cyclohexa-1,5-dien-1-yloxy)trimethylsilane) is silyl enol ether of cyclohexenone
  • The diels-alder reactions with dienophiles α-acetoxyacrylonitrile, acrylonitrile and α-chloroacrylonitrile has been studied.2-(trimethylsiloxy)-1,3-cyclohexadiene may be used as a diene in the synthesis of isoquinuclidinone 2-aza[2.2.2]octa-3,5-dione by reacting with p-toluenesulfonyl cyanide

Specifications

CAS 59084-06-9
Molecular Formula C10H13N3O2
Linear Formula C10H13N3O2
MDL Number MFCD00040728
Quantity 5 mL
Molecular Weight (g/mol) 207.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.