Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Nitrophenyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. N11321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N11321G 1 g
N11325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N11321G Supplier TCI America Supplier No. N11321G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3205-25-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD02683380
InChI Key DSDBYQDNNWCLHL-UHFFFAOYNA-N
Synonym 2-Nitro-alpha-methylbenzyl Alcohol
PubChem CID 97851
IUPAC Name 1-(2-nitrophenyl)ethan-1-ol
SMILES CC(O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Boiling Point 107°C
Quantity 1 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Nitrophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.