Learn More
Ace Glass, Inc. PTFE insert, 1/2" NPT, #11 compression fitting9 to 10.5mm OD tubing
SDP
Catalog No. 50977908
Encompass
Compression fitting PTFE inserts for 1/2NPT to #11 Ace-Thred. Fits 9 to 10.5mm O.D. tubing. For use with ACE PTFE reaction vessel heads.
Catalog No. 50-977-908 Supplier Ace Glass, Inc. Supplier No. 1287111
Compression fitting PTFE inserts for 1/2NPT to #11 Ace-Thred. Fits 9 to 10.5mm O.D. tubing. For use with ACE PTFE reaction vessel heads.