Learn More
Ace Glass, Inc. PTFE insert, 1/2" NPT, #7 compression fitting6.5 to 7mm OD tubing
SDP
Catalog No. 50977907
Encompass
Compression fitting PTFE inserts for 1/2NPT to #7 Ace-Thred. Fits 6.5 to 7mm O.D. tubing. For use with ACE PTFE reaction vessel heads.
Catalog No. 50-977-907 Supplier Ace Glass, Inc. Supplier No. 1287107
Compression fitting PTFE inserts for 1/2NPT to #7 Ace-Thred. Fits 6.5 to 7mm O.D. tubing. For use with ACE PTFE reaction vessel heads.