Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-O-Isopropylidene-alpha-D-glucurono-6,3-lactone 98.0+%, TCI America™
SDP

Catalog No. I06881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06881G Supplier TCI America Supplier No. I06881G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 20513-98-8
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00061641
InChI Key BDBGJSXZKMTMGP-BOFVIFSBSA-N
Synonym d-glucurono-6,3-lactone acetonide
PubChem CID 134129430
IUPAC Name (3aS,3bS,6S,6aS,7aR)-6-hydroxy-2,2-dimethyl-3b,6,6a,7a-tetrahydro-3aH-furo[2,3]furo[2,4-b][1,3]dioxol-5-one
SMILES CC1(OC2C3C(C(C(=O)O3)O)OC2O1)C

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 1 g
Formula Weight 216.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-O-Isopropylidene-alpha-D-glucurono-6,3-lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.