Learn More
Chemglass Life Sciences 1/2" OD Stainless Steel Support Rod, Bent 90°
SDP
Catalog No. CG349803
Encompass_Preferred
1/2" OD S.S. Support Rod, Bent 90¿. For use with the CG-3498 series of Temperature Monitors. CG-3498-03 1/2¿ OD rod, bent 90¿ is necessary when supporting a temperature monitor or controller to a reactor stand or laboratory hood lattice.
Catalog No. CG349803 Supplier Chemglass Life Sciences Supplier No. CG349803
1/2" OD S.S. Support Rod, Bent 90¿. For use with the CG-3498 series of Temperature Monitors. CG-3498-03 1/2¿ OD rod, bent 90¿ is necessary when supporting a temperature monitor or controller to a reactor stand or laboratory hood lattice.