Learn More
THORLABS INC 1/2" OPTICAL POST,SS, 8-32,5/P
Catalog No. NC2185895
Encompass
1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 3", 5 Pack
This item is not returnable. View return policy
Catalog No. NC2185895 Supplier THORLABS INC Supplier No. TR3P5
1/2" Optical Post, SS, 8-32 Setscrew, 1/4"-20 Tap, L = 3", 5 Pack