Learn More
Medchemexpress LLC 1-(2-oxo-2-(3'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl)ethyl)-5-(pyrrolidin-1-ylsulfonyl)pyridin-2(1H)-one | 1996629-79-8 | 99.6% | 490.49 g/mol | C24H21F3N2O4S | 10MG
SDP
Catalog No. 5000236109
Encompass_Preferred
TBOPP is a research-grade small-molecule inhibitor selective for DOCK1, intended for preclinical and biochemical studies. It inhibits DOCK1 with an IC50 of 8.4 μM and binds the DOCK1 DHR-2 domain with a Kd of 7.1 μM; TBOPP has reported anti-tumor activity in multiple tumor models. The compound is supplied as a white to off-white solid with high purity and is typically prepared in DMSO for solution-based assays.
Catalog No. 50-002-36109 Supplier Medchemexpress LLC Supplier No. HY12471110MG
TBOPP is a research-grade small-molecule inhibitor selective for DOCK1, intended for preclinical and biochemical studies. It inhibits DOCK1 with an IC50 of 8.4 μM and binds the DOCK1 DHR-2 domain with a Kd of 7.1 μM; TBOPP has reported anti-tumor activity in multiple tumor models. The compound is supplied as a white to off-white solid with high purity and is typically prepared in DMSO for solution-based assays.