Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Pentanediol

Catalog No. 260282250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-028-2250ML 250 mL
26-028-250ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-028-2250ML Supplier Sigma Aldrich Supplier No. 260282250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Pentanediol

Specifications

CAS 5343-92-0
Density 0.971 g/mL (at 25°C)
Boiling Point 206°C
Molecular Formula C5H12O2
Refractive Index n20/D 1.439 (literature)
Linear Formula CH3CH2CH2CH(OH)CH2OH
Quantity 250 mL
MDL Number MFCD00010736
Molecular Weight (g/mol) 104.15
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.