Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-pentyl)-piperazine

Catalog No. CDS004840 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS004840 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS004840 Supplier Sigma Aldrich Supplier No. CDS004840100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-pentyn-2-ol, a secondary homopropargylic alcohol, is toxic with lc50 value of 160.4-pentyn-2-ol was used in the synthesis of new fischer-type rhenium (iii) benzoyldiazenido-2-oxacyclocarbenes
  • It was also used in the preparation of c-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide

Specifications

CAS 82499-96-5
Molecular Formula C9H20N2
Linear Formula C9H20N2
MDL Number MFCD02093553
Quantity 25 g
Molecular Weight (g/mol) 156.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.