Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Phenyl-1,3-thiazol-4-yl)ethan-1-one, 97%, Thermo Scientific™

Catalog No. SPB03431DA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB03431DA 1 g
SPB03431EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPB03431DA Supplier Thermo Scientific Chemicals Supplier No. SPB03431DA
Only null left
Add to Cart

Chemical Identifiers

CAS 10045-52-0
Molecular Formula C11H9NOS
Molecular Weight (g/mol) 203.259
MDL Number MFCD00068112
InChI Key ZOOGZFPRAKXWKI-UHFFFAOYSA-N
Synonym 1-2-phenylthiazol-4-yl ethanone, 1-2-phenyl-1,3-thiazol-4-yl ethan-1-one, 1-2-phenyl-1,3-thiazol-4-yl ethanone, 4-acetyl-2-phenylthiazole, 1-2-phenyl-1,3-thiazol-4-yl-1-ethanone, ethanone, 1-2-phenyl-4-thiazolyl, 4-acetyl-2-phenyl-1,3-thiazole, maybridge1_008631, buttpark 15357-18, 2-phenyl-4-acetylthiazole
PubChem CID 604819
IUPAC Name 1-(2-phenyl-1,3-thiazol-4-yl)ethanone
SMILES CC(=O)C1=CSC(=N1)C2=CC=CC=C2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 203.26
Percent Purity 95%
Chemical Name or Material 1-(2-Phenyl-1, 3-thiazol-4-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.